Learn More
Thermo Scientific™ Hypersil™ SAS C1 LC Columns Available on GSA/VA Contract for Federal Government customers only.
Separate moderately polar and multi-functional compounds with Thermo Scientific™ Hypersil™ SAS C1 LC columns.
Manufacturer:  Thermo Scientific™ 30505254630
 View more versions of this product
Catalog No. 03-050-540
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Separate moderately polar and multi-functional compounds with Thermo Scientific™ Hypersil™ SAS C1 LC columns.
Classical Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
  • A short alkyl chain reversed phase material
  • Selectivity for polar and multifunctional compounds
  • Can be used in ion-pair separations
Ordering Information:
Other column dimensions are available. Also available in 10µm. Please call Customer Service for more information.
Description & Specifications
Specifications
3%
Reversed Phase
Yes
400bar
5μm
120Å
60°C
C1
Analytical Column
4.6mm
250mm
Silica, Spherical Fully Porous Type A
2 to 8
170m²/g
L13