Learn More
Thermo Scientific™ Hypersil™ Silica Drop-in Guard Cartridges Available on GSA/VA Contract for Federal Government customers only.
Convenience, economy and effective protection for extending column lifetime.
Manufacturer:  Thermo Scientific™ 30005012101
 View more versions of this product
Catalog No. 03-150-520
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ Hypersil Silica Drop-in Guard Cartridges offer convenience, economy and effective protection for extending column lifetime. Available for all popular Thermo Scientific phases.
  • The 10mm design offers maximum protection with minimal increase in retention Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Available in packs of 4
Requires:
Cartridge holder