Learn More
Thermo Scientific™ Hypo-Vial Septa
Thermo Scientific™ Hypo-Vial Septa are ideal to use with borosilicate glass sample bottles for heating and autoclaving.
Specifications
For Use With borosilicate glass sample bottles for heating and autoclaving
View More Specs
Products 1
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
PI13237
View Documents
Thermo Scientific™
TS13237
Silicone This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ Hypo-Vial Septa are ideal to use with borosilicate glass sample bottles for heating and autoclaving.
  • Closure for 50 and 125mL glass sample bottles
  • Hycar and silicone plug-type septa and flat PTFE/Silicone laminated discs (sold separately)
  • Fits 6-125mL Hypo-Vial sample storage vials
  • Enable syringe-needle access to vial contents
Specifications
Specifications
borosilicate glass sample bottles for heating and autoclaving