Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hypophosphorous acid, Acros Organics™

Click to view available options
Quantity:
1L
Packaging:
Plastic bottle
6303-21-5,7732-18-5
HO2P
63.98
GQZXNSPRSGFJLY-UHFFFAOYSA-N
hypophosphite ion,hypophosphorus acid,phosphinic acid, ion 1-,hydroxyphosphanone,oxo-,e5-phosphanol,hydroxidooxidophosphorus,perfluoroalkyl c6-c12 phosphonic acid,diperfluoroalkyl c6-c12 phosphinic acid,hydroxy-,e5-phosphanone,perfluoro-c6-c12 ethyl hydrogen phosphate
22497
CHEBI:36363
phosphenous acid
OP=O
6303-21-5,7732-18-5
HO2P
63.98
GQZXNSPRSGFJLY-UHFFFAOYSA-N
hypophosphite ion,hypophosphorus acid,phosphinic acid, ion 1-,hydroxyphosphanone,oxo-,e5-phosphanol,hydroxidooxidophosphorus,perfluoroalkyl c6-c12 phosphonic acid,diperfluoroalkyl c6-c12 phosphinic acid,hydroxy-,e5-phosphanone,perfluoro-c6-c12 ethyl hydrogen phosphate
22497
CHEBI:36363
phosphenous acid
OP=O
Boiling Point 108.0°C (1013.0hPa),108.0°C (1013.0 hPa)
Melting Point -25.0°C,−25.0°C
Linear Formula H3PO2
Solubility Information Solubility in water: soluble. Other solubilities: miscible with ethanol, immiscible with hydrocarbons and halogenated, hydrocarbons
Formula Weight 66
Density 1.2700g/mL
Packaging Plastic bottle
Quantity 1L
Chemical Name or Material Hypophosphorous acid