Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hypromellose, Substitution Type 2208, 100,000 mPa.s, USP, 4-12%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9004-65-3
C56H108O30
1261.45
PUSNGFYSTWMJSK-UHFFFAOYNA-N
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O
9004-65-3
C56H108O30
1261.45
PUSNGFYSTWMJSK-UHFFFAOYNA-N
1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O

H1393, 9004-65-3

Spectrum™ Hypromellose, Substitution Type 2208, 100,000 mPa.s, USP is an artificial tear used to relieve dry eye conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.015
Percent Purity 4 to 12%
Loss on Drying 0.05
Packaging Poly Bottle
Quantity 125 g