Learn More
Thermo Scientific™ HyPURITY™ C8 Guard Cartridges
Thermo Scientific™ HyPURITY™ C8 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
Manufacturer:  Thermo Scientific™ 22205014001
 View more versions of this product
Catalog No. 22205014001
Add to cart
Description
Description
Thermo Scientific™ HyPURITY™ C8 Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • High quality, highly pure silica columns
Requires:
Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.
Specifications
Specifications
Uniguard Direct-connection and Stand Alone Holder
L7
5µm
2 - 9
10mm
8 %
Yes
190Å
200m²/g
4mm
Documents
Accessories