Learn More
Sklar InstrumentsSupplier Diversity Partner Z-Type Hysterectomy Forceps, Sklar, 12 in., Serrated Tips, Curved, Longitudinally Grooved, Flared Shanks, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 902798
Z-Type Hysterectomy Forceps, Sklar, 12 in., Serrated Tips, Curved, Longitudinally Grooved, Flared Shanks, Stainless Steel. Sklar Z-Type Hysterectomy Forceps are a ratcheted, finger ring forceps used in vaginal and abdominal hysterectomy procedures. The handles of these forceps are offset so as not to obstruct the physician's view of the surgical site. They are available in 4 lengths with various curvatures to meet the needs of the procedure, physician preference, and patient comfort. This product has strongly curved jaws with serrated tips, and longitudinal grooves. It is 12 inches long.
Catalog No. 50-143-6594