Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aspergillus Rabbit anti-Fungi, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29826
Description
Rabbit polyclonal antibody raised against Aspergillus.
- High specificity
- High reactivity
Specifications
Aspergillus | |
Polyclonal | |
Rabbit polyclonal antibody raised against Aspergillus. | |
In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
Aspergillus species extracts. | |
500 μL | |
Primary | |
Fungi | |
Liquid |
Immunohistochemistry | |
Unconjugated | |
Immunohistochemistry The optimal working dilution should be determined by the end user. | |
Rabbit | |
IgG purification | |
RUO | |
This antibody reacts with soluble proteins of Common Aspergillus species. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction