Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bacillus Rabbit anti-Bacteria, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29827
Description
Rabbit polyclonal antibody raised against Bacillus.
- High specificity
- High reactivity
Specifications
Bacillus | |
Polyclonal | |
Rabbit polyclonal antibody raised against Bacillus. | |
In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
Purified spores of Bacillus cereus and Bacillus subtilis. | |
500 μL | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot | |
Unconjugated | |
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user. | |
Rabbit | |
IgG purification | |
RUO | |
Bacteria | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction