Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

I-BET 151 hydrochloride, Tocris Bioscience™

Catalog Number p-4817046
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
C23H21N5O3
7-(3,5-Dimethyl-4-isoxazolyl)-1,3-dihydroxy-8-methoxy-1-[(1R)-1-(2-pyridinyl)ethyl]-2H-imidazo[4,5-c]quinolin-2-one hydrochloride
This item is not returnable. View return policy
C23H21N5O3
7-(3,5-Dimethyl-4-isoxazolyl)-1,3-dihydroxy-8-methoxy-1-[(1R)-1-(2-pyridinyl)ethyl]-2H-imidazo[4,5-c]quinolin-2-one hydrochloride

BET bromodomain inhibitor; also promotes differentiation of hiPSCs into megakaryocytes

BET bromodomain inhibitor; blocks recruitment of BET to chromatin. Induces apoptosis and G0/G1 cell cycle arrest in MLL-fusion leukemic cell lines in vitro (IC50 values are 15, 26, 120 and 192 nM for NOMO1, MV4;11, MOLM13 and RS4;11 cell lines respectively); reduces BCL2 expression in NOMO1 cells. Improves survival in two rodent models of MLL-fusion leukemia in vivo. Enhances differentiation of human iPSC into megakaryocytes.
Quantity 10 mg
Formula Weight 451.9
Percent Purity >98%
Chemical Name or Material I-BET 151 hydrochloride
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit