Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Borrelia burgdorferi dog IgG/IgM ELISA Kit

Enzyme-linked immunosorbent assay for the detection of dog IgG and IgM antibodies to Borrelia burgdorferi.

Manufacturer:  Abnova™ KA4895

Catalog No. 89-032-343


Add to cart

Description

Description

Borrelia burgdorferi dog IgG/IgM ELISA Kit is an enzyme-linked immunosorbent assay for the detection of dog IgG and IgM antibodies to Borrelia burgdorferi.
Specifications

Specifications

HRP
Qualitative
Borrelia burgdorferi dog IgG/IgM ELISA Kit
100 μL (Prediluted)
Dog
1 Kit
Serum
Borrelia burgdorferi dog IgG/IgM ELISA Kit is an enzyme-linked immunosorbent assay for the detection of dog IgG and IgM antibodies to Borrelia burgdorferi.
Conjugate Diluent 2: 13mL
For research use only (RUO)
Store the kit at 4°C. Do not freeze.
Colorimetric
Canine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit