Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ I-BRD9 Available on GSA/VA Contract for Federal Government customers only.

Potent and selective BRD9 inhibitor

$222.50 - $931.50

Specifications

Chemical Name or Material 5-Ethyl-4,5-dihydro-4-oxo-N-(tetrahydro-1,1-dioxido-2H-thiopyran-4-yl)-7-[3-(trifluoromethyl)phenyl]thieno[3,2-c]pyridine-2-carboximidamide
Molecular Formula C22H22F3N3O3S2
CAS 1714146-59-4
Product Type Target: Bromodomain Inhibitors
Formula Weight 497.55
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
559110 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5591/10
10mg Each for $222.50
Add to cart
 
559150 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5591/50
50mg Each for $931.50
Add to cart
 
Description

Description

Potent and selective BRD9 inhibitor (pIC50 = 7.3). Exhibits >70-fold selectivity for BRD9 over a panel of 34 other bromodomains and >700-fold selectivity over the BET family. Downregulates CLEC1, DUSP6, FES and SAMSN1 genes in Kasumi-1 cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

5-Ethyl-4,5-dihydro-4-oxo-N-(tetrahydro-1,1-dioxido-2H-thiopyran-4-yl)-7-[3-(trifluoromethyl)phenyl]thieno[3,2-c]pyridine-2-carboximidamide
1714146-59-4
497.55
C22H22F3N3O3S2
Target: Bromodomain Inhibitors
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit