Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-CBP 112, Tocris Bioscience™
Click to view available options
Quantity:
10mg
50mg
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity | 10mg |
Formula Weight | 468.59 |
Percent Purity | >98% |
Chemical Name or Material | I-CBP 112 |
Safety and Handling
Recommended Storage : Store at +4°C