Learn More
Chlamydia trachomatis Goat anti-Bacteria, Polyclonal, Abnova™
Goat Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29831
Catalog No. 89-033-676
Add to cart
Description
Description
Goat polyclonal antibody raised against Chlamydia trachomatis.
  • High specificity
  • High reactivity
Specifications
Specifications
Chlamydia trachomatis
Unconjugated
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Chlamydia trachomatis serovar groups.
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Bacteria
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow
Goat polyclonal antibody raised against Chlamydia trachomatis.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Goat
IgG purification
RUO
Primary
This antibody reacts with disrupted and purified elementary bodies.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)