Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Chlamydia trachomatis IgG ELISA Kit

Enzyme immunoassay for the detection of IgG antibodies to Chlamydia trachomatis in human serum or plasma

Manufacturer:  Abnova™ KA4826

Catalog No. 89-032-280


Add to cart

Description

Description

Intended for detection of specific IgG antibodies in a sample by means of a sandwich type of the EIA method (i.e. a solid phase coated with specific antigen – antibody from the analyzed sample – labeled antibody).
Specifications

Specifications

5 to 320 U/mL
Qualitative; Quantification
Chlamydia trachomatis IgG ELISA Kit
100 μL
Human
1 Kit
Plasma, Serum
Chlamydia trachomatis IgG ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgG antibodies to Chlamydia trachomatis.
Microtitre Plate: 96 wells, Negative Control (Calibrator 1): 5 U/mL (2mL), CUT-OFF (Calibrator 2): 20 U/mL (3mL), Positive Control (Calibrator 3): 80 U/mL (2mL), Calibrator 4: 320 U/mL (2mL), Conjugate: 15mL, Sample Diluent 2: 105mL, TMB-Complete 2: 15mL,
For research use only (RUO)
Store the kit at 4°C. Do not freeze.
Colorimetric
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit