Learn More
Clostridium glutamate dehydrogenase Mouse anti-Clostridium Species, Clone: HM177, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13503
Catalog No. 89-033-468
Add to cart
Description
Description
Mouse monoclonal antibody raised against Clostridium glutamate dehydrogenase (GDH).
  • High specificity
  • High reactivity
Specifications
Specifications
Clostridium glutamate dehydrogenase
HM177
Mouse monoclonal antibody raised against Clostridium glutamate dehydrogenase (GDH).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with recombinant and native GDH of Clostridium difficile.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG1
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Clostridium Species
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)