Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ I.D. PRO™ Plus, LS2000 and BradyMarker™ XC Plus Printer Labels
For use with I.D. PRO Plus, LS2000 and BradyMarker XC Plus Printers. Brady™ wire and cable marking labels are made from nylon cloth with a permanent adhesive.
Supplier: Brady™ WML911499
Description
- For marking wires and cables
- Available in a variety of sizes
Specifications
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) | |
Acrylic, Permanent | |
Roll | |
Adhesive wire marking label | |
I.D. PRO Plus, LS2000 and BradyMarker XC Plus Wiremarker Printers | |
1 x 1.25 in. (W x H) |
Compatible surface: rough, smooth and textured | |
White | |
Nylon Cloth (B-499) | |
Wire and cable marking, panel identification | |
250/Roll | |
0.0065 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction