Brady™ I.D. PRO™ Plus, LS2000 and BradyMarker™ XC Plus Printer Labels

For use with I.D. PRO Plus, LS2000 and BradyMarker XC Plus Printers. Brady™ wire and cable marking labels are made from nylon cloth with a permanent adhesive.

Manufacturer: Brady™ WML517499

 View more versions of this product

Catalog No. 19-109-451

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    For Use With (Equipment) I.D. PRO Plus, LS2000 and BradyMarker XC Plus Wiremarker Printers
    Adhesive Type Permanent Acrylic
    Quantity 250/Roll
    Description Compatible surface: rough, smooth and textured
    Format Roll
    For Use With (Application) Wire and cable marking, panel identification
    Label Size (English) W x H: 1.5 x 0.75 in.
    Item Description Abrasion, Chemical, Humidity, Low Temperature, UV and Weather-Resistant
    Material B-499 Nylon cloth
    Product Type Adhesive wire marking label
    Color White
    Thickness (English) 0.0065 in.
    Certifications/Compliance UL Recognized; CSA Accepted; Halogen Free (DIN VDE 0472/part 815)

    • For marking wires and cables
    • Available in a variety of sizes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit