Learn More
Drosophila Medium and Vials Products for Science Education
Easy to use.
$11.01
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
S22321A Products for Science Education
View Documents
Biological Resource Center
10 VIALS
Culture Vials
Pack of 10 for $11.01
Add to cart
Description
Description
Drosophila Medium
  • Media is mold resistant and ready to use — combine equal volumes of Drosophila medium and cold tap water, sprinkle dried yeast on surface, let it stand for one minute, and you are ready to inoculate
  • Blue food coloring provides a contrasting background for observing pale larvae
  • Each 1 gal. bag makes 300 vials of medium
Drosophila Culture Vials with Plugs
  • Disposable transparent plastic containers with plastic foam plugs
  • Sold in packs of 10