Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-E[k] Mouse anti-Mouse, BV786, Clone: 14-4-4S, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744681
Description
I-E[k] Monoclonal antibody specifically detects I-E[k] in Mouse samples. It is validated for Flow Cytometry.Specifications
I-E[k] | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
14-4-4S | |
Brilliant Violet 786 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction