Learn More
Escherichia coli O157 Mouse anti-E. coli, Clone: HM017, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13545
Catalog No. 89-033-508
Add to cart
Description
Description
Mouse monoclonal antibody raised against Escherichia coli O157.
  • High specificity
  • High reactivity
Specifications
Specifications
Escherichia coli O157
HM017
Mouse monoclonal antibody raised against Escherichia coli O157.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with O157 of Escherichia coli.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG2b
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
E. coli
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)