Learn More
Giardia lamblia Mouse anti-Parasite, Clone: HM014, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13551
Catalog No. 89-033-514
Add to cart
Description
Description
Mouse monoclonal antibody raised against Giardia lamblia.
  • High specificity
  • High reactivity
Specifications
Specifications
Giardia lamblia
HM014
Mouse monoclonal antibody raised against Giardia lamblia.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with antigen of Giardia lamblia.
ELISA, Immunofluorescence, Lateral Flow, Western Blot
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG2b
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Parasite
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)