Learn More
Helicobacter pylori Rabbit anti-Helicobacter pylori, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29847
Catalog No. 89-033-692
Add to cart
Description
Description
Rabbit polyclonal antibody raised against Helicobacter pylori.
  • High specificity
  • High reactivity
Specifications
Specifications
Helicobacter pylori
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Helicobacter pylori ATCC strain 43504.
RUO
Primary
This antibody reacts with all antigens of Helicobacter pylori.
ELISA, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against Helicobacter pylori.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Rabbit
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Helicobacter pylori
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)