Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB zeta Rat anti-Mouse, PerCP-eFluor™ 710, Clone: LK2NAP, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 46680180

 View more versions of this product

Catalog No. 50-112-3881


Add to cart

Description

Description

Description: The LK2NAP monoclonal antibody reacts with mouse IkB zeta (INAP, MAIL). IkBz is a member of the IkB family of cytoplasmic proteins and transcription factors and also shares structural homology with Bcl-3. It is induced in monocytes, macrophages and B cells in response to TLR and IL-1R signaling. In NK cells, IkBz is rapidly upregulated in response to IL-12 and IL-18 and is necessary for production of IFN gamma. In T cells, IkBz is not necessary for IFN gamma and IL-4 production by Th1 and Th2 cells, respectively. However in Th17 cells, IkBz acts in concert with RORa and RORgt to upregulate IL-17A, IL-17F, IL-21, IL-22 and IL-23R expression. Applications Reported: This LK2NAP antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This LK2NAP antibody has been tested by intracellular staining and flow cytometric analysis of LPS-activated mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

PerCP-eFluor® 710 can be used in place of PE-Cy5, PE- Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 µm post-manufacturing filtered. I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombit IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.
Specifications

Specifications

IkB zeta
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9EST8
FLJ30225, FLJ34463, IKBZ, INAP, MAIL
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
LK2NAP
PerCP-eFluor™ 710
Nfkbiz
Liquid
Nfkbiz
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
80859
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.