Learn More
Listeria monocytogenes Mouse anti-Bacteria, Clone: HM218, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13609
Catalog No. 89-033-572
Add to cart
Description
Description
Mouse monoclonal antibody raised against Listeria monocytogenes.
  • High specificity
  • High reactivity
Specifications
Specifications
Listeria monocytogenes
HM218
Mouse monoclonal antibody raised against Listeria monocytogenes.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bacteria
ELISA, Immunofluorescence
Unconjugated
ELISA Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
IgG2b
RUO
Primary
This antibody reacts with p60 of Listeria monocytogenes.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)