Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Mycoplasma pneumoniae IgG ELISA Kit
Enzyme-linked immunosorbent assay for the detection of IgG antibodies to Mycoplasma pneumoniae.
Supplier: Abnova™ KA4849
Description
Intended for detection of specific IgG antibodies in a sample by means of a sandwich type of the EIA method (i.e. a solid phase coated with specific antigen – antibody from the analyzed sample – labeled antibody).Specifications
5 to 320 U/mL | |
HRP | |
Mycoplasma pneumoniae IgG ELISA Kit | |
Human | |
Microtitre Plate: 96 wells, Negative Control (Calibrator 1): 5 U/mL (2mL), CUT-OFF (Calibrator 2): 20 U/mL (3mL), Positive Control (Calibrator 3): 80 U/mL (3mL), Calibrator 4: 320 U/mL (2mL), Conjugate: 15mL, Sample Diluent 2: 105mL, TMB-Complete 2: 15mL, Wash Solution: 20x concentrated buffer: 75mL, Stop Solution: 1 M sulphuric acid solution: 15mL | |
For research use only (RUO) | |
Store the kit at 4°C. Do not freeze. |
Mycoplasma pneumoniae IgG ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgG antibodies to Mycoplasma pneumoniae. | |
Colorimetric | |
Human Plasma, Human Serum | |
Qualitative; Quantification | |
1 Kit | |
100 μL | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction