Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neisseria gonorrhoeae Rabbit anti-Bacteria, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29800
Description
Rabbit polyclonal antibody raised against Neisseria gonorrhoeae .
- High specificity
- High reactivity
Specifications
Neisseria gonorrhoeae | |
Polyclonal | |
Rabbit polyclonal antibody raised against Neisseria gonorrhoeae . | |
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide). | |
Neisseria gonorrhoeae whole cells. | |
RUO | |
This antibody reacts with all Neisseria gonorrhoeae antigens. May react with related microbes. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot | |
Unconjugated | |
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user. | |
Rabbit | |
500 μL | |
Primary | |
Bacteria | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction