Learn More
Masterflex™ I/P™ Precision Pump Tubing, PharMed™ BPT
Protect your light-sensitive samples
$281.50 - $593.50
Specifications
Color Opaque Beige
Autoclavable Autoclavable
Material PharMed BPT
View More Specs
Products 3
Catalog Number Tubing Size Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Tubing Size Diameter (Metric) Inner Price Quantity & Availability  
13-200-305
View Documents
26 6.4 mm
Pack for $281.50
Add to cart
13-310-148
View Documents
73 9.5 mm
Pack for $495.50
Add to cart
13-310-155
View Documents
82 12.7 mm
Pack for $593.50
Add to cart
Description
Description
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Specifications
Specifications
Opaque Beige
Autoclavable
PharMed BPT