Learn More
Pichia pastoris Rabbit anti-Yeast, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29804
Catalog No. 89-033-649
Add to cart
Description
Description
Rabbit polyclonal antibody raised against Pichia pastoris.
  • High specificity
  • High reactivity
Specifications
Specifications
Pichia pastoris
Unconjugated
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Liquid
Pichia pastoris whole cells.
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Yeast
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot
Rabbit polyclonal antibody raised against Pichia pastoris.
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Rabbit
IgG
RUO
Primary
This antibody reacts with all antigens of Pichia pastoris. Strong reactivity with C. albicans and weak reactivity with S. cerevisiae.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)