Learn More
Salmonella LPS core Mouse anti-Salmonella, Clone: HM038, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13647
Catalog No. 89-033-610
Add to cart
Description
Description
Mouse monoclonal antibody raised against Salmonella LPS core.
  • High specificity
  • High reactivity
Specifications
Specifications
Salmonella LPS core
HM038
Mouse monoclonal antibody raised against Salmonella LPS core.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with LPS core of Salmonella.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG2a
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Salmonella
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)