Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Streptococcus agalactiaeserotype III, ATCC™ BAA-22™†

For culture purposes and QC applications.

$79.50 - $210.00

Specifications

ATCC® BAA-22
Target Organism Class Bacteria
Specimen Type Serotype III
Format KWIK-STIK™
For Use With (Application) Culture purpose and QC application
View More Specs

Products 3
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
23-017-279
View Documents Promotion Details
Microbiologics™
01234K
KWIK-STIK™ 6/Pk.
Pack of 6 for $210.00
Add to cart
 
23-017-280
View Documents Promotion Details
Microbiologics™
01234P
KWIK-STIK™ 2/Pk.
Pack of 2 for $79.50
Add to cart
 
23-017-281
View Documents Promotion Details
Microbiologics™
01234L
LYFO DISK™ 6/Pk.
Pack of 6 for $179.30
Add to cart
 
Description

Description

KWIK-STIK™
  • Gives accurate, repeatable results
  • Microorganism, hydrating fluid and inoculating swab all in one self-contained device
  • Each device features single microorganism strain in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
  • For added convenience, peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Escherichia coli
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
Specifications

Specifications

BAA-22
Serotype III
Culture purpose and QC application
2
Bacteria
KWIK-STIK™
Lyophilized Organism
Streptococcus agalactiae
Safety and Handling

Safety and Handling

SDS
Product Certifications

† The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit