Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus group A Rabbit anti-Streptococcus, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

$346.00

Specifications

Antigen Streptococcus group A
Host Species Rabbit
Target Species Streptococcus
Applications ELISA
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-033-659 View Documents Abnova Corporation
PAB29814
1 mg Each for $346.00
Add to cart
 
89-033-661 View Documents Abnova Corporation
PAB29816
500 ug Each for $346.00
Add to cart
 
Description

Description

Rabbit polyclonal antibody raised against Streptococcus group A.

  • High specificity
  • High reactivity
Specifications

Specifications

Streptococcus group A
Streptococcus
RUO
Liquid
Primary
In 0.02 M sodium phosphate, 0.15 M sodium chloride (0.09% sodium azide).
Rabbit
ELISA
Unconjugated
Store at 4°C.
Polyclonal

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit