Learn More
Streptococcus pneumoniae Mouse anti-Streptococcus, Clone: HM047, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13656
Catalog No. 89-033-619
Add to cart
Description
Description
Mouse monoclonal antibody raised against Streptococcus pneumoniae.
  • High specificity
  • High reactivity
Specifications
Specifications
Streptococcus pneumoniae
HM047
Mouse monoclonal antibody raised against Streptococcus pneumoniae.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with C-polysaccharide (phosphorylcholine epitope) of Streptococcus pneumoniae.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG3
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Streptococcus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)