Learn More
Toxoplasma gondii Mouse anti-Protozoa, Clone: HM161, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13660
Catalog No. 89-033-623
Add to cart
Description
Description
Mouse monoclonal antibody raised against Toxoplasma gondii.
  • High specificity
  • High reactivity
Specifications
Specifications
Toxoplasma gondii
HM161
Mouse monoclonal antibody raised against Toxoplasma gondii.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
IgG2a
RUO
Primary
This antibody reacts with SAG1 protein of Toxoplasma gondii.
ELISA, Immunofluorescence, Western Blot
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Toxoplasma gondii extracts.
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Protozoa
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)