Learn More
CERILLIANT™ Human Insulin Solution, 100 μg/mL in PBS, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco I0340.1ML
Catalog No. 01-001-502
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Human insulin is a peptide hormone, composed of 51 amino acids, produced by beta cells of the pancreatic islets. Insulin regulates the metabolism of carbohydrates, fats and protein by promoting the absorption of, especially glucose from the blood into liver, fat and skeletal muscle cells. Insulin levels are routinely measured to evaluate insulin production; diagnose insulin-producing tumors; determine hypoglycemia or to identify insulin resistance. This certified Snap-N-Spike™ solution of the fully-intact human insulin protein is suitable for use as starting material in calibrators and controls in LC-MS/MS insulin testing methods.
Specifications
Specifications
11061-68-0
5807.57
Ampule
-20°C
100 μg/mL in PBS
C257H383N65O77S6
0.1 mL
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation