Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IA-6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541650
Description
Peptide sequence: KRTGGLYRVR LAERVFPLLG PQGAPPFLEE APSASLPGAE RATPPTREEP Sequence homology: Human: 100%.
IA-6, also known as insulinoma-associated 2 or INSM2, is related to IA-1 (INSM1), a zinc-finger factor that is expressed in the developing nervous system. While the exact function of IA-6 is not known, other related insulinoma-associated proteins such as IA-2 are major autoantigens in type 1 diabetes. The homology exhibited by IA-6 to IA-1 has led to speculation that IA-6 may also play a role in the regulation of gene transcription.
Specifications
IA-6 | |
Polyclonal | |
Unconjugated | |
INSM2 | |
IA6; IA-6; INSM transcriptional repressor 2; Insm2; INSM2 variant 1; insulinoma-associated 2; insulinoma-associated protein 2; insulinoma-associated protein IA-6; methylated in liver tumor 1; mlt 1; mlt1; Nbla106; Zinc finger protein IA-6 | |
Rabbit | |
Protein A | |
RUO | |
84684 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96T92 | |
INSM2 | |
Synthetic peptide directed towards the N-terminal of human INSM2 (aa 11-60). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction