Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IAH1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548275

Catalog No. PA548275


Add to cart

Description

Description

The function of this protein remains unknown.
Specifications

Specifications

IAH1
Polyclonal
PBS with 0.09% sodium azide
Q9DB29
Isoamyl acetate-hydrolyzing esterase 1 homolog, 31--, IAH1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
67732
Western Blot
Unconjugated
Iah1
Liquid
IAH1
KLH conjugated synthetic peptide between 78-106 amino acids from the Central region of human IAH1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit