Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IARS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP184637
Description
IARS2 Polyclonal specifically detects IARS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IARS2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
EC 6.1.1, EC 6.1.1.5, FLJ10326, ILERS, isoleucine tRNA ligase 2, mitochondrial, Isoleucine--tRNA ligase, isoleucine-tRNA synthetase 2, mitochondrial, isoleucyl-tRNA synthetase 2, mitochondrial, isoleucyl-tRNA synthetase, mitochondrial, mitochondrial isoleucine tRNA synthetase | |
Rabbit | |
Affinity Purified | |
RUO | |
55699 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
IARS2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ALNGMVEMMDRRPYWCISRQRVWGVPIPVFHHKTKDEYLINSQTTEHIVKLVEQHGSDIWWTLPPEQLLPKEVLSEVGGPDALEYVPGQD | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction