Learn More
IARS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549883
Catalog No. PA549883
Add to cart
Description
Description
The antibody detects endogenous levels of total IARS2 protein.
IARS2 belongs to the class-I aminoacyl-tRNA synthetase family. Members of class I have two highly conserved sequence motifs. They aminoacylate at the 2 ft.-OH of an adenosine nucleotide, and are usually monomeric or dimeric (one or two subunits, respectively). Both classes of aminoacyl-tRNA synthetases are multidomain proteins. The catalytic domains of all the aaRSs of a given class are found to be homologous to one another, while class I and class II aaRSs are unrelated to one another. The class I aaRSs have the ubiquitous Rossmann fold and have the antiparallel beta-strands architecture while the class II aaRSs have a unique fold made up of antiparallel beta-strands.
Specifications
Specifications
IARS2
Polyclonal
Unconjugated
IARS2
Liquid
IARS2
Synthesized peptide derived from internal of human IARS2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NSE4, Q8BIJ6
EC 6.1.1.5, isoleucine-tRNA synthetase 2, mitochondrial,
Rabbit
IgG
100 μL
-20°C
Primary
381314, 55699