Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IARS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549566

Catalog No. PA549566

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAS, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of isoleucine-tRNA synthetase exist, a cytoplasmic form and a mitochondrial form. This gene encodes the mitochondrial isoleucine-tRNA synthetase which belongs to the class-I aminoacyl-tRNA synthetase family. [provided by RefSeq, Dec 2014]
Specifications

Specifications

IARS2
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NSE4, Q8BIJ6
Isoleucine--tRNA ligase, mitochondrial, Isoleucyl-tRNA synthetase, IleRS, IARS2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
381314, 55699
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
IARS2
Liquid
IARS2
synthetic peptide corresponding to 85-113aa of human SYIM.
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit