Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IB-MECA, Tocris Bioscience™

Catalog Number p-4783521
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
152918-18-8
C18H19IN6O4
510.292
HUJXGQILHAUCCV-MOROJQBDSA-N
ib-meca, 3-ib-meca, piclidenoson, n 6-ibamu, n 6-3-iodobenzyl-5'-n-methylcarboxamidoadenosine, unii-30679umi0n, n 6-3-iodobenzyl adenosine-5'-n-methyluronamide, 1-deoxy-1-6-3-iodophenyl methyl amino-9h-purin-9-yl-n-methyl-beta-d-ribofuranuronamide, 3-iodobenzyl-5'-n-methylcarboxamidoadenosine, n 6-3-iodo-benzyl adenosine-5'-n-methyluronamide
123683
CHEBI:73286
(2S,3S,4R,5R)-3,4-dihydroxy-5-[6-[(3-iodophenyl)methylamino]purin-9-yl]-N-methyloxolane-2-carboxamide
CNC(=O)C1C(C(C(O1)N2C=NC3=C2N=CN=C3NCC4=CC(=CC=C4)I)O)O
This item is not returnable. View return policy
152918-18-8
C18H19IN6O4
510.292
HUJXGQILHAUCCV-MOROJQBDSA-N
ib-meca, 3-ib-meca, piclidenoson, n 6-ibamu, n 6-3-iodobenzyl-5'-n-methylcarboxamidoadenosine, unii-30679umi0n, n 6-3-iodobenzyl adenosine-5'-n-methyluronamide, 1-deoxy-1-6-3-iodophenyl methyl amino-9h-purin-9-yl-n-methyl-beta-d-ribofuranuronamide, 3-iodobenzyl-5'-n-methylcarboxamidoadenosine, n 6-3-iodo-benzyl adenosine-5'-n-methyluronamide
123683
CHEBI:73286
(2S,3S,4R,5R)-3,4-dihydroxy-5-[6-[(3-iodophenyl)methylamino]purin-9-yl]-N-methyloxolane-2-carboxamide
CNC(=O)C1C(C(C(O1)N2C=NC3=C2N=CN=C3NCC4=CC(=CC=C4)I)O)O

A3 selective agonist

Potent and selective A3 adenosine receptor agonist (Ki values are 1.1, 54 and 56 nM for A3, A1 and A2A receptors respectively). Cardioprotective, reduces infarct size upon reperfusion in rats.
Quantity 5 mg
Formula Weight 510.29
Percent Purity >97%
Chemical Name or Material IB-MECA
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit