Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ IB™ Model Serum Filters

Catalog No. 0268162 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
2.75 in.
4 in.
6 in.
Length (Metric):
70mm
100mm
152mm
Diameter (English) Outside:
0.5 in.
0.6 in.
Diameter (Metric) Outside:
13mm
16mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Diameter (English) Outside Diameter (Metric) Outside
02-681-62 2.75 in. 70mm 0.6 in. 16mm
02-681-66 4 in. 100mm 0.5 in. 13mm
02-681-65 2.75 in. 70mm 0.5 in. 13mm
02-681-63 4 in. 100mm 0.6 in. 16mm
02-681-64 6 in. 152mm 0.6 in. 16mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-681-62
Only null left
Add to Cart
Add to Cart

Store serum up to seven days when it is essential to prevent any exchange of ions.

  • One-way valve permits a physical gap to be created between the serum/plasma and clot, and prevents backflow into the primary collection tube
  • Use with glass or plastic collection tubes
  • Technique reduces second centrifugation, tube labeling and sample preparation time
  • Allows direct sampling into automatic laboratory equipment
  • Eliminates fibrin, particulate matter and equipment downtime

Specifications

For Use With (Equipment) Glass or Plastic collection tubes
Diameter (English) Outside 0.6 in.
Length (Metric) 70mm
For Use With (Application) Eliminates fibrin, particulate matter and equipment downtime
Length (English) 2.75 in.
Material Polyethylene
Product Type Serum Filter
Diameter (Metric) Outside 16mm
Quantity 100 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.