Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec IB Series Portable Cantilever Storage Rack Available on GSA/VA Contract for Federal Government customers only.

A versatile economical means of vertical storage of multiple cadavers that adjusts easily for a wide variety of heights. Mopec™ IB Series Portable Cantilever Storage Rack is an adjustable cantilever racks that is designed to support up to 500 pounds per tier.

Manufacturer:  Mopec IB100

 View more versions of this product

Catalog No. 22-444-145


Add to cart

Description

Description

  • All stainless steel construction
  • Adjustable arms
  • 4 heavy-duty locking casters
  • Maximum weight of 500 lbs. per arm (226.79kg)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

1 Year Warranty

Specifications

Specifications

Stainless Steel
4
110.54cm
78.74cm
171.45cm
Holds up to 226.79kg
Cadaver Storage Racks
43.52 in.
31 in.
67.5 in.
Holds up to 500lbs
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit