Learn More
Iba1/AIF1 Rabbit anti-Human,Mouse, KHL, Clone: 16B14, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB117725UL
 View more versions of this product
Catalog No. ZRB117725MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Iba1/AIF1 Monoclonal antibody specifically detects Iba1/AIF1 in Human,Mouse samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Iba1/AIF1
16B14
KHL
Purified
AIF1, G!, IBA1
KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human Allograft inflammatory factor 1 (AIF1).
Target molecular weight 16.70 kDa calculated; ∼16 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_001614.3
Allograft inflammatory factor 1;Ionized calcium-binding adapter molecule 1;Protein G1
Rabbit
IgG
Protein A Purified
Signaling
Primary
Human, Mouse