Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBC 293, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
306935-41-1
C10H11N3O2
205.217
RUTVRAJKELSHCC-UHFFFAOYSA-N
1-isopropyl-1h-1,2,3-benzotriazole-5-carboxylic acid,1-isopropyl-1h-benzo d 1,2,3 triazole-5-carboxylic acid,1-isopropyl-1,2,3-benzotriazole-5-carboxylic acid,1-isopropyl-1h-benzotriazole-5-carboxylic acid,1h-benzotriazole-5-carboxylic acid, 1-1-methylethyl,1-isopropylbenzotriazole-5-carboxylic acid,1h-benzotriazole-5-carboxylicacid, 1-1-methylethyl,1-propan-2-yl-1h-1,2,3-benzotriazole-5-carboxylic acid,1-methylethyl benzotriazole-5-carboxylic acid,maybridge1_005244
2736690
1-propan-2-ylbenzotriazole-5-carboxylic acid
CC(C)N1C2=C(C=C(C=C2)C(=O)O)N=N1
This item is not returnable. View return policy
306935-41-1
C10H11N3O2
205.217
RUTVRAJKELSHCC-UHFFFAOYSA-N
1-isopropyl-1h-1,2,3-benzotriazole-5-carboxylic acid,1-isopropyl-1h-benzo d 1,2,3 triazole-5-carboxylic acid,1-isopropyl-1,2,3-benzotriazole-5-carboxylic acid,1-isopropyl-1h-benzotriazole-5-carboxylic acid,1h-benzotriazole-5-carboxylic acid, 1-1-methylethyl,1-isopropylbenzotriazole-5-carboxylic acid,1h-benzotriazole-5-carboxylicacid, 1-1-methylethyl,1-propan-2-yl-1h-1,2,3-benzotriazole-5-carboxylic acid,1-methylethyl benzotriazole-5-carboxylic acid,maybridge1_005244
2736690
1-propan-2-ylbenzotriazole-5-carboxylic acid
CC(C)N1C2=C(C=C(C=C2)C(=O)O)N=N1

Selective agonist for the GPR109B (HM74) receptor. Antilipolytic

Highly selective agonist for GPR109B (HM74), human orphan G-protein-coupled receptor expressed in adipocytes. Selective for GPR109B over niacin receptor GPR109A (HM74A); EC 50 values are 400 nM and > 1 μM respectively. Inhibits isoproterenol-stimulated lipolysis in vitro in isolated human adipocytes.
Quantity 50mg
Formula Weight 205.22
Percent Purity >99%
Chemical Name or Material IBC 293

Recommended Storage : Store at Room Temperature