Learn More
IBI Scientific™ Low Melting Point Agarose
Designed for preparative DNA and RNA electrophoresis, cloning of tissue culture cells and viral plague assays, and fragment recovery following electrophoresis. IBI Scientific™ Low Melting Point Agarose has lower gelling point and melting point when compared to Standard Agarose allows quick and thorough digestion during extraction procedures and easy in-gel manipulations.
Manufacturer:  IBI Scientific™ IB70059
 View more versions of this product
Catalog No. 15-455-205
  Check Availability
Add to cart
Specifications
Description
  • Lower gelling and melting temperature than standard agarose
  • Lower gel strength than standard agarose while still easy to handle
  • Higher gel transparency than standard agarose
  • Greater sieving capacity than standard agarose
  • Separates and finely resolves molecules >1000bp
  • DNase- and RNase-Free
Description & Specifications
Specifications
Used as gel matrix and preparation of large DNA samples for plugs
1kg