Learn More
IBI Scientific™ TUNAIR™ Shake Flask System
Provides optimum growth conditions for aerobic microorganisms, mammalian cells and plant cells
$19.37 - $550.00
Products 31
Catalog Number Mfr. No. Product Type No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Product Type No. per Pack Price Quantity & Availability  
15-453-302
View Documents
IBI Scientific™
SS2001C
Shake Flask System Kit; Working vol. 100mL; No-Baffle; w Dri-Gauze lining and slip on cap Each
Pack for $386.50
Add to cart
15-453-304
View Documents
IBI Scientific™
SS2002C
Shake Flask System Kit; working vol. 100mL; Half-Baffle; w Dri-Gauze lining and slip on cap Each
Pack for $234.00
Add to cart
15-453-303
View Documents
IBI Scientific™
SS2001S
Shake Flask System Kit; working vol. 100mL; No-Baffle; w silicone lining and slip on cap Each
Pack for $524.50
Add to cart
15-453-305
View Documents
IBI Scientific™
SS2002S
Shake Flask System Kit; working vol. 100mL; Half-Baffle; w silicone lining and slip on cap Each
Pack for $439.00
Add to cart
15-453-306
View Documents
IBI Scientific™
SS2003C
Shake Flask System Kit; Working vol.: 100mL; Full-Baffle; w Dri-Gauze lining and slip on cap Each
Pack for $276.00
Add to cart
15-453-307
View Documents
IBI Scientific™
SS2003S
Shake Flask System Kit; Working vol.: 100mL; Full-Baffle; w silicone lining and slip on cap Each
Pack for $463.00
Add to cart
15-453-308
View Documents
IBI Scientific™
SS2011
Shake Flask; No-Baffle (normal throw); 300mL; Working vol.: 100mL Each
Pack for $34.85
Add to cart
15-453-309
View Documents
IBI Scientific™
SS2012
Shake Flask; Half-Baffle (vortex motion); 300mL; Working vol.: 100mL Each
Pack for $34.85
Add to cart
15-453-310
View Documents
IBI Scientific™
SS2013
Shake Flask; Full-Baffle (propeller motion); 300mL; Working vol.: 100mL Each
Pack for $34.85
Add to cart
15-453-318
View Documents
IBI Scientific™
SS4002
Shake Flask; Half-Baffle (vortex motion); 300mL; Working vol.: 100mL 4
Pack for $73.50
Add to cart
15-453-322
View Documents
IBI Scientific™
SS5013
Shake Flask; Full-Baffle (propeller motion); 2.5L; Working vol.: 1L Each
Pack for $60.60
Add to cart
15-453-317
View Documents
IBI Scientific™
SS4001
Shake Flask; No-Baffle (normal throw); 300mL; Working vol.: 100mL 4
Pack for $108.80
Add to cart
15-453-320
View Documents
IBI Scientific™
SS5011
Shake Flask; No-Baffle (normal throw); 2.5L; Working vol.: 1L Each
Pack for $58.40
Add to cart
15-453-321
View Documents
IBI Scientific™
SS5012
Shake Flask; Half-Baffle (vortex motion); 2.5L; Working vol.: 1L Each
Pack for $55.20
Add to cart
15-453-319
View Documents
IBI Scientific™
SS4003
Shake Flask; Full-Baffle (propeller motion); 300mL; Working vol.: 100mL 4
Pack for $88.40
Add to cart
15-453-323
View Documents
IBI Scientific™
SS6001C
Shake Flask System Kit; Working vol.: 1L; No-Baffle Each
Pack for $388.50
Add to cart
15-453-325
View Documents
IBI Scientific™
SS6002C
Shake Flask System Kit; Working vol.: 1L; Half-Baffle Each
Pack for $211.00
Add to cart
15-453-328
View Documents
IBI Scientific™
SS6003S
Shake Flask System Kit; Working vol.: 1L; Full-Baffle Each
Pack for $277.00
Add to cart
15-453-330
View Documents
IBI Scientific™
SS8002
Shake Flask; Half-Baffle (vortex motion); 2.5L; Working vol.: 1L 4
Pack for $128.00
Add to cart
15-453-329
View Documents
IBI Scientific™
SS8001
Shake Flask; No-Baffle (normal throw); 2.5L; Working vol.: 1L 4
Pack for $128.00
Add to cart
15-453-324
View Documents
IBI Scientific™
SS6001S
Shake Flask System Kit; Working vol.: 1L; No-Baffle; With silicone lining and slip on cap Each
Pack for $550.00
Add to cart
15-453-326
View Documents
IBI Scientific™
SS6002S
Shake Flask System Kit; Working vol.: 1L; Half-Baffle; w/ silicone lining and slip on cap Each
Pack for $312.00
Add to cart
15-453-327
View Documents
IBI Scientific™
SS6003C
Shake Flask System Kit; Working vol.: 1L; Full-Baffle; w/ Dri-Gauze lining and slip on cap Each
Pack for $211.00
Add to cart
15-453-331
View Documents
IBI Scientific™
SS8003
Shake Flask; Full-Baffle (propeller motion); 2.5L; Working vol.: 1L 4
Pack for $148.00
Add to cart
15-453-332
View Documents
IBI Scientific™
SS7001
Shake Flask System Kit 1
Pack for $276.00
Add to cart
15-453-312
View Documents
IBI Scientific™
SS3015
Two-piece cap; For 2.5L flask 1
Pack for $30.15
Add to cart
15-453-311
View Documents
IBI Scientific™
SS3014
Two-piece cap; For 300mL flask 1
Pack for $19.37
Add to cart
15-453-314
View Documents
IBI Scientific™
SS3017
Silicone membrane; For 2.5L flask 5
Pack for $149.80
Add to cart
15-453-313
View Documents
IBI Scientific™
SS3016
Silicone membrane; For 300mL flask 5
Pack for $104.00
Add to cart
15-453-315
View Documents
IBI Scientific™
SS3018
Dri-gauze; For 300mL flask 5
Pack for $37.50
Add to cart
15-453-316
View Documents
IBI Scientific™
SS3019
Dri-gauze; For 2.5L flask 5
Pack for $47.40
Add to cart
Description
Description
  • Three flask design based on applications: No-Baffle (normal throw), Half-Baffle (vortex motion), and Full-Baffle (propeller motion)
  • Increased oxygen absorption rate due to unique baffling and turbo-vane closure design
  • Silicone or nitrocellulose filter linings are 0.22μ
  • Slip on two piece polypropylene cap eliminates need of flaming and protect neck of flask from airborne particles
  • Autoclavable
Applications:
Microbiology and biotechnology