Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abnova™ IBRDC2 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00255488T01
Catalog No. 89-948-179
Specifications
Specifications
293T
BC063311.1
pCMV-IBRDC2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
IBRDC2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 33.33kDa
Transient overexpression cell lysate was tested with anti-IBRDC2 antibody (Mfr. No. H00255488-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human