Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen, 99%, Thermo Scientific Chemicals

Catalog Number p-4510328
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
15687-27-1
C13H18O2
206.29
MFCD00010393
HEFNNWSXXWATRW-UHFFFAOYNA-N
3672
CHEBI:5855
2-[4-(2-methylpropyl)phenyl]propanoic acid
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
15687-27-1
C13H18O2
206.29
MFCD00010393
HEFNNWSXXWATRW-UHFFFAOYNA-N
3672
CHEBI:5855
2-[4-(2-methylpropyl)phenyl]propanoic acid
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Ibuprofen, 2-4-isobutylphenyl propanoic acid, CAS # 15687-27-1, is a laboratory form of the over-the-counter nonsteroidal anti-inflammatory (NSAID) drug.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Ibuprofen (2-4-isobutylphenyl propanoic acid) is a common NSAID that reduces pyrogen release and inflammation
  • Ibuprofen is a racemic mixture that consists of an R-enantiomer and S-enantiomer; the S-enantiomer is proposed to be the stronger acting agent
  • It is a propionic acid derivative, a short-chain saturated fatty acid bound to ethane
  • Ibuprofen inhibits cyclooxygenases and reduces prostaglandin formation, inhibiting fever and pain

Applications

  • Ibuprofen reduces both local and systemic inflammation; suitable for use in vitro or in vivo to study specific or broad mechanisms
  • Intended for sensitive biochemical applications such as HPLC
TRUSTED_SUSTAINABILITY
Melting Point 75.0°C to 78.0°C
Color White
Boiling Point 157.0°C (4.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2C6H4CH(CH3)COOH
Quantity 10 g
Merck Index 15,4919
Synonym ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
Solubility Information Solubility in water: insoluble. Other solubilities: readily soluble in most organic solvents
Formula Weight 206.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ibuprofen
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 239-784-6

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit