Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ICAM-1/CD54 Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795865PLATES

 View more versions of this product

Catalog No. NB010212


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse ICAM-1/CD54.
Specifications

Specifications

HRP
Sandwich ELISA
8.75 pg/mL
3383
RUO
ICAM-1/CD54
Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers
Mouse
ICAM1
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 560 pg/mL is recommended.
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse ICAM-1/CD54.
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 20 ng of recombinant Mouse ICAM-1/CD54.
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse ICAM-1/CD54 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-ICAM-1/CD54 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit
    5 Plates
    8.75-560 pg/mL
    BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit